Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanesulfonic acid sodium salt, monohydrate; 99.9%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15181305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15181305 5 g
ICN15181325 25 g
ICN15181380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15181305 Supplier MP Biomedicals, Inc Supplier No. 0215181305

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Item Name 1-Pentanesulfonic Acid, Sodium Salt, Monohydrate
Content And Storage Store at Room Temperature.
Form Crystalline Powder
Quantity 5 g
Appearance White
Molecular Formula C5H11NaO3S
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES [Na+].CCCCCS([O-])(=O)=O
IUPAC Name sodium pentane-1-sulfonate
Molecular Weight (g/mol) 174.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.