Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanol, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018350
Click to view available options
Quantity:
1 mL
5 mL
10 mL

1-Pentanol is a type of alcohol which has been proposed as an alternate for conventional gasoline and diesel fuels.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 71-41-0
Vapor Pressure 1 mmHg (13.6°C)
Vapor Density 3 (vs. air)
Density 0.811 g/mL (at 25°C)
Flash Point 47°C
Physical Form Neat
Linear Formula CH3(CH2)4 OH
MDL Number MFCD00002977
Melting Point -78°C
Boiling Point 136°C to 138°C
Synonym n-Amyl alcohol; Pentyl alcohol
Type Analytical Standard
Quantity 10 mL
Formula Weight 88.15
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.