Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanol, ACS, 99+%

Catalog No. p-7049902
Click to view available options
Quantity:
250 mL
1 L
4 L

Filtered through 0.2μ filters. Suitable for spectrophotometry, chromatography | CAS: 71-41-0 | C5H12O | 88.15 g/mol

1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Melting Point -79°C
Color Colorless
Density 0.812 g/mL
Boiling Point 136°C to 138°C
Flash Point 49°C (120°F)
Odor Mild
Refractive Index 1.4103
Linear Formula CH3(CH2)4OH
Quantity 250 mL
UN Number UN1105
Beilstein 1730975
Merck Index 14,7118
Solubility Information Miscible with water,alcohol,ether,acetone and most organic solvents.
Formula Weight 88.15
Percent Purity ≥99%
Grade ACS
Physical Form Liquid
Chemical Name or Material 1-Pentanol
Show More Show Less
Hazard Category H226-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H332-H315-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
EINECSNumber 200-752-1
RTECSNumber SB9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.