Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Pentanol, Reagent, ACS, 98%, Spectrum™ Chemical

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
Description
Spectrum™ Chemical 1-Pentanol, Reagent, ACS
Specifications
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 4 x 4 L |
UN Number | UN1105 |
Residue after Evaporation | 0.003% |
Percent Purity | 98% |
Grade | ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction