Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanol, ReagentPlus™, ≥99%, Solstice

Catalog No. 60044825 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-825 2.5 L Glass Bottle
60-044-824 1 L Glass Bottle
60-044-826 20 L Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-044-825 Supplier Solstice Supplier No. 1389752.5L

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Melting Point -78°C
Color Colorless
Density 0.811g/mL at 25°C
Boiling Point 136°C to 138°C
Flash Point 49°C (120.2°F)
Packaging Glass Bottle
Refractive Index n20/D 1.409(lit.)
Linear Formula CH3(CH2)4OH
Quantity 2.5 L
UN Number UN1105
Beilstein 1730975
Formula Weight 88.15g/mol
Percent Purity ≥99%
Grade ReagentPlus
Chemical Name or Material 1-Pentanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338-P370 + P378
Precautionary Statement H226-H315-H319-H332-H335
DOTInformation class 3 - PG 3 - Pentanols
EINECSNumber 200-752-1
RTECSNumber SB9800000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.