Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanol, ReagentPlus™, ≥99%, Honeywell™

Catalog No. p-7120037
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Melting Point -78°C
Color Colorless
Density 0.811g/mL at 25°C
Boiling Point 136°C to 138°C
Flash Point 49°C (120.2°F)
Packaging Glass Bottle
Refractive Index n20/D 1.409(lit.)
Linear Formula CH3(CH2)4OH
Quantity 1 L
UN Number UN1105
Beilstein 1730975
Formula Weight 88.15g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Chemical Name or Material 1-Pentanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338-P370 + P378
Precautionary Statement H226-H315-H319-H332-H335
DOTInformation class 3 - PG 3 - Pentanols
EINECSNumber 200-752-1
RTECSNumber SB9800000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.