Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Penten-3-ol, 97%

Catalog No. p-179614
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 616-25-1 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-25-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00004573
InChI Key VHVMXWZXFBOANQ-UHFFFAOYNA-N
Synonym 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
PubChem CID 12020
IUPAC Name pent-1-en-3-ol
SMILES CCC(O)C=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Beige
Density 0.8390g/mL
Boiling Point 114.0°C to 115.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4220 to 1.4260
Linear Formula C2H5CH(OH)CH=CH2
Quantity 5 g
Beilstein 01,443
Specific Gravity 0.839
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 86.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Penten-3-ol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 210-472-1
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.