Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Pentyl isocyanate, 97%
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.878 |
| Boiling Point | 136°C to 137°C |
| Flash Point | 32°C (89°F) |
| Odor | Pungent |
| Refractive Index | 1.415 |
| Linear Formula | CH3(CH2)4NCO |
| Quantity | 5 g |
| UN Number | UN3080 |
| Beilstein | 1746688 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H312+H332-H315-H318-H334-H335 |
| Hazard Statement | GHS H Statement H301-H330-H334-H226-H315-H319-H335 Toxic if swallowed. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P342+P311-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction