Learn More
1-Pentylamine, 98%
Description
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.
Solubility
Soluble in alcohol, ether.
Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | -55°C |
Density | 0.751 |
Boiling Point | 103°C to 105°C |
Flash Point | −1°C (30°F) |
Odor | Amine-like |
Refractive Index | 1.411 |
Quantity | 500 g |
UN Number | UN1106 |
Beilstein | 505953 |
Sensitivity | Air Sensitive |
Show More |
Safety and Handling
Hazard Category | H225-H302+H312-H314-H331-H335 |
Hazard Statement | GHS H Statement H225-H331-H314-H318-H302-H312 Highly flammable liquid and vapor. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES |
EINECSNumber | 203-780-2 |
RTECSNumber | SC0300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.