Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentylamine, 98%

Catalog No. p-7081626
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 110-58-7 | C5H13N | 87.166 g/mol

1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

Solubility
Soluble in alcohol, ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-58-7
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008236
InChI Key DPBLXKKOBLCELK-UHFFFAOYSA-N
Synonym amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
PubChem CID 8060
ChEBI CHEBI:74848
IUPAC Name pentan-1-amine
SMILES CCCCCN

Specifications

Melting Point -55°C
Density 0.751
Boiling Point 103°C to 105°C
Flash Point −1°C (30°F)
Odor Amine-like
Refractive Index 1.411
Quantity 25 g
UN Number UN1106
Beilstein 505953
Sensitivity Air Sensitive
Merck Index 14,598
Solubility Information Soluble in alcohol,ether.
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material 1-Pentylamine
Show More Show Less
Hazard Category H225-H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H225-H331-H314-H318-H302-H312
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES
EINECSNumber 203-780-2
RTECSNumber SC0300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.