Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1,2-propanedione-2-oxime, 99%

Catalog No. p-183282
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle

CAS: 119-51-7 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-51-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00002115
InChI Key YPINLRNGSGGJJT-JXMROGBWSA-N
Synonym 1-phenyl-1,2-propanedione-2-oxime, 2-hydroxyimino propiophenone, isonitrosopropiophenone, 2-hydroxyiminopropiophenone, 2-hydroxyimino-1-phenylpropan-1-one, propiophenone, isonitroso, 2-isonitrosopropiophenone, alpha-oximinopropiophenone, 1-phenyl-1,2-propanedione 2-oxime, .alpha.-oximinopropiophenone
PubChem CID 9566063
IUPAC Name (2E)-2-hydroxyimino-1-phenylpropan-1-one
SMILES C\C(=N/O)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 111.0°C to 115.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5COC(=NOH)CH3
Quantity 10 g
Beilstein 07, III, 3464
Solubility Information Solubility in water: 1.3g/L (20°C)
Formula Weight 163.18
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material 1-Phenyl-1, 2-propanedione-2-oxime, 99%
Show More Show Less
EINECSNumber 204-329-2
RTECSNumber UH2975000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.