Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1,2-propanedione, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
579-07-7
C9H8O2
148.16
MFCD00008755
BVQVLAIMHVDZEL-UHFFFAOYSA-N
1-phenyl-1,2-propanedione,acetylbenzoyl,acetyl benzoyl,benzoylacetyl,methylphenylglyoxal,pyruvophenone,1,2-propanedione, 1-phenyl,phenylmethyldiketone,benzoyl methyl ketone,methyl phenyl diketone
11363
CHEBI:63552
1-phenylpropane-1,2-dione
CC(=O)C(=O)C1=CC=CC=C1
579-07-7
C9H8O2
148.16
MFCD00008755
BVQVLAIMHVDZEL-UHFFFAOYSA-N
1-phenyl-1,2-propanedione,acetylbenzoyl,acetyl benzoyl,benzoylacetyl,methylphenylglyoxal,pyruvophenone,1,2-propanedione, 1-phenyl,phenylmethyldiketone,benzoyl methyl ketone,methyl phenyl diketone
11363
CHEBI:63552
1-phenylpropane-1,2-dione
CC(=O)C(=O)C1=CC=CC=C1
Specific Gravity 1.1
Refractive Index 1.5300 to 1.5340
Boiling Point 103.0°C to 105.0°C (14.0 mmHg)
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Density 1.1000g/mL
Flash Point 84°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5COCOCH3
Beilstein 07, 677
Solubility Information Solubility in water: insoluble
Formula Weight 148.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Phenyl-1, 2-propanedione, 98%
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 209-435-2

TSCA : TSCA