Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1,3-butanedione 98.0+%, TCI America™
SDP

Catalog No. P0160100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0160100G 100 g
P016025G 25 g
P0160500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0160100G Supplier TCI America Supplier No. P0160100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93-91-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00008786
InChI Key CVBUKMMMRLOKQR-UHFFFAOYSA-N
Synonym benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
PubChem CID 7166
IUPAC Name 1-phenylbutane-1,3-dione
SMILES CC(=O)CC(=O)C1=CC=CC=C1

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 262°C
Quantity 100 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-1,3-butanedione
EINECSNumber (3)-3494
RTECSNumber EK3540200
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.