Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-cyclohexene 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
771-98-2
C12H14
158.24
MFCD00001542
WCMSFBRREKZZFL-UHFFFAOYSA-N
1-phenyl-1-cyclohexene,1-phenylcyclohexene,1-phenylcyclohex-1-ene,benzene, 1-cyclohexen-1-yl,phenylcyclohexene,cyclohexenylbenzene,benzene, cyclohexenyl,2,3,4,5-tetrahydro-1,1'-biphenyl,cyclohexene, 1-phenyl,trans-1-phenylcyclohexene
13043
cyclohexen-1-ylbenzene
C1CCC(=CC1)C2=CC=CC=C2
771-98-2
C12H14
158.24
MFCD00001542
WCMSFBRREKZZFL-UHFFFAOYSA-N
1-phenyl-1-cyclohexene,1-phenylcyclohexene,1-phenylcyclohex-1-ene,benzene, 1-cyclohexen-1-yl,phenylcyclohexene,cyclohexenylbenzene,benzene, cyclohexenyl,2,3,4,5-tetrahydro-1,1'-biphenyl,cyclohexene, 1-phenyl,trans-1-phenylcyclohexene
13043
cyclohexen-1-ylbenzene
C1CCC(=CC1)C2=CC=CC=C2
Density 0.9900g/mL
Refractive Index 1.5680 to 1.57
Boiling Point 251°C to 253°C
Melting Point -11°C
Flash Point 103°C
Specific Gravity 0.99
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula C6H5C6H9
Beilstein 05, II, 419
Formula Weight 158.24
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 1-Phenyl-1-cyclohexene, 95%
Show More Show Less

EINECSNumber : 212-242-6

RTECSNumber : GW6600000

TSCA : TSCA