Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-cyclopentanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. P148725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P148725G 25 g
P14875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P148725G Supplier TCI America Supplier No. P148725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77-55-4
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00001372
InChI Key RHPCYZLXNNRRMB-UHFFFAOYSA-N
Synonym 1-phenylcyclopentanecarboxylic acid, 1-phenyl-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopentanecarboxylic acid, 1-phenylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-phenyl, pubchem22923, acmc-209pao, phenylcyclopentanecarboxylic acid, phenylcyclopentane carboxylic acid
PubChem CID 66167
IUPAC Name 1-phenylcyclopentane-1-carboxylic acid
SMILES OC(=O)C1(CCCC1)C1=CC=CC=C1

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 25 g
Formula Weight 190.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-1-cyclopentanecarboxylic Acid
EINECSNumber (9)-1186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.