Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-propyne, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
673-32-5
C9H8
116.16
MFCD00009272
GHUURDQYRGVEHX-UHFFFAOYSA-N
1-phenyl-1-propyne,prop-1-yn-1-ylbenzene,benzene, 1-propynyl,methylphenylacetylene,2-propynylbenzene,1-phenylpropyne,1-phenylpropyne-1,unii-448376bfqc,prop-1-ynyl-benzene,1-methyl-2-phenylacetylene
69601
prop-1-ynylbenzene
CC#CC1=CC=CC=C1
673-32-5
C9H8
116.16
MFCD00009272
GHUURDQYRGVEHX-UHFFFAOYSA-N
1-phenyl-1-propyne,prop-1-yn-1-ylbenzene,benzene, 1-propynyl,methylphenylacetylene,2-propynylbenzene,1-phenylpropyne,1-phenylpropyne-1,unii-448376bfqc,prop-1-ynyl-benzene,1-methyl-2-phenylacetylene
69601
prop-1-ynylbenzene
CC#CC1=CC=CC=C1
Specific Gravity 0.92
Boiling Point 185.0°C
Packaging Glass bottle
Density 0.9200g/mL
Flash Point 62°C
Color Yellow
Refractive Index 1.5630 to 1.5650
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5C≡CCH3
Beilstein 05, 514
Formula Weight 116.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1-Phenyl-1-propyne
Show More Show Less

EINECSNumber : 211-607-7