Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(+/-)-1-Phenylethylamine, 98+%
Click to view available options
Quantity:
100 mL
500 mL
Description
Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.
Solubility
Soluble in water at 20°C 4.2 g/L.
Notes
Air sensitive. Store away from air, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.
Solubility
Soluble in water at 20°C 4.2 g/L.
Notes
Air sensitive. Store away from air, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Specifications
Specifications
| Density | 0.945 |
| Boiling Point | 69°C to 70°C (10 mmHg) |
| Flash Point | 79°C (174°F) |
| Odor | Odorless |
| Refractive Index | 1.527 |
| Quantity | 100 mL |
| UN Number | UN2735 |
| Beilstein | 636127 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,6026 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Combustible liquid. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 210-545-8 |
| RTECSNumber | DP5775000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction