Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenylimidazole, 97%, Thermo Scientific™

Catalog No. AC308780010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 7164-98-9
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00041204
InChI Key SEULWJSKCVACTH-UHFFFAOYSA-N
Synonym 1-phenyl-1h-imidazole, phenylimidazole, n-phenylimidazole, 1h-imidazole, 1-phenyl, 1h-imidazole, phenyl, n-phenyl imidazole, pubchem12611, acmc-20aos8, 1-phenylimidazole, 1-phenyl-1h-imidazole #
PubChem CID 81595
IUPAC Name 1-phenyl-1H-imidazole
SMILES C1=CN(C=N1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 13°C
Color Yellow
Density 1.1400g/mL
Boiling Point 142°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5990 to 1.601
Quantity 1 g
Specific Gravity 1.14
Formula Weight 144.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Phenylimidazole, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.