Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyloctane, 99%

Catalog No. p-3779444
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

CAS: 2189-60-8 | C14H22 | 190.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2189-60-8
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00009564
InChI Key CDKDZKXSXLNROY-UHFFFAOYSA-N
Synonym n-octylbenzene, 1-phenyloctane, benzene, octyl, phenyloctane, octane, 1-phenyl, octyl-benzene, 1-octylbenzene, octyl benzene, 4-octylbenzene, acmc-1cplg
PubChem CID 16607
IUPAC Name octylbenzene
SMILES CCCCCCCCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -36°C
Color Colorless
Density 0.8500g/mL
Boiling Point 261°C to 263°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.485
Linear Formula C6H5(CH2)7CH3
Quantity 25 mL
Beilstein 05, 453
Specific Gravity 0.85
Formula Weight 190.33
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Phenyloctane
Show More Show Less
EINECSNumber 218-582-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.