Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenylvinylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. p-7035499
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143825-84-7
Molecular Formula C14H19BO2
Molecular Weight (g/mol) 230.114
MDL Number MFCD06659923
InChI Key RMGBWPMWUZSIMH-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-1-phenylvinyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl, 1-phenylvinylboronic acid pinacolester, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11031671
IUPAC Name 4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C(=C)C2=CC=CC=C2

Specifications

Melting Point 40°C to 42°C
Quantity 250 mg
Formula Weight 230.11
Percent Purity 95%
Chemical Name or Material 1-Phenylvinylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.