Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Propanol, for spectroscopy ACS
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy

Specifications
Specifications
Melting Point | -127.0°C |
Density | 0.8000g/mL |
Boiling Point | 97.0°C |
Flash Point | 15°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Refractive Index | 1.3840 to 1.3860 |
Linear Formula | CH3CH2CH2OH |
Quantity | 1 L |
Residue after Evaporation | 0.001% max. |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause drowsiness or dizziness. Causes serious eye damage. Highly flammable liquid and vapor. |
Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physici |
EINECSNumber | 200-746-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction