Alfa Aesar™ 1-Propylamine, 99+%

Manufacturer: Alfa Aesar™ 36635K2

Quantity 1L

 View more versions of this product

Catalog No. AA36635K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107-10-8
    Molecular Formula CH3CH2CH2NH2
    Formula Weight 59.11
    MDL Number MFCD00008205
    UN Number UN1277
    Beilstein 1098243
    Merck Index 14,7843
    Synonym 1-Aminopropane
    Chemical Name or Material 1-Propylamine

    Specifications

    Formula Weight 59.11
    Assay Percent Range >99+%
    Molecular Formula CH3CH2CH2NH2
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1277
    Synonym 1-Aminopropane
    Melting Point −83°C
    Health Hazard 1 Danger
    Flash Point −12° (10°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPYLAMINE
    TSCA TSCA
    Refractive Index 1.389
    Physical Form Liquid
    MDL Number MFCD00008205
    Merck Index 14,7843
    Density 0.719g/mL
    pH 12.6
    Odor Amine-like
    CAS 107-10-8
    Beilstein 1098243
    Boiling Point 48° to 49°C
    Health Hazard 2 GHS H Statement
    H225-H311-H314-H318-H302
    Highly flammable liquid and vapor.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    RTECS Number UH9100000
    Sensitivity Air Sensitive
    Quantity 1L
    Color Colorless
    Chemical Name or Material 1-Propylamine
    Assay ≥99%

    GHS H Statement
    H225-H311-H314-H318-H302
    Highly flammable liquid and vapor.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit