Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propylamine, 99+%

Catalog No. AA36635AK
Click to view available options
Quantity:
250 mL
1 L

Catalog No. AA36635AK

Supplier: Thermo Scientific Chemicals 036635AK

Only null left
Add to Cart

CAS: 107-10-8 | C3H9N | 59.112 g/mol

1-Propylamine is used in chemical analysis and to make other chemicals. It is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Also it can be used as petroleum additive and preservative. It is used in the production of fiber and engineering plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-10-8
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.112
MDL Number MFCD00008205
InChI Key WGYKZJWCGVVSQN-UHFFFAOYSA-N
Synonym propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
PubChem CID 7852
ChEBI CHEBI:39870
IUPAC Name propan-1-amine
SMILES CCCN

Specifications

Melting Point °C to 83°C
Density 0.719 g/mL
Boiling Point 48°C to 49°C
Flash Point -12°C (10°F)
Assay Percent Range >99+%
Refractive Index 1.389
Quantity 250 mL
UN Number UN1277
Beilstein 1098243
Sensitivity Air Sensitive
Merck Index 14,7843
Solubility Information Fully miscible with water. Highly soluble in ethanol and acetone,soluble in benzene and chloroform and slightly soluble in carbon tetrachloride.
Formula Weight 59.11
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material 1-Propylamine
Show More Show Less
Hazard Category H225-H290-H302-H311+H331-H314-H335
Hazard Statement GHS H Statement
H225-H311-H314-H318-H302
Precautionary Statement P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPYLAMINE
EINECSNumber 203-462-3
RTECSNumber UH9100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.