Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyreneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P162525G
Click to view available options
Quantity:
5 g
25 g

Specifications

CAS 164461-18-1
Melting Point 251°C
Molecular Formula C16H11BO2
Quantity 25 g
Synonym 1-pyrenylboronic acid, 1-pyreneboronic acid, pyrene-1-boronic acid, 1-boronopyrene, 1-pyrenyl boronic acid
InChI Key MWEKPLLMFXIZOC-UHFFFAOYSA-N
SMILES B(C1=C2C=CC3=CC=CC4=C3C2=C(C=C1)C=C4)(O)O
IUPAC Name pyren-1-ylboronic acid
Molecular Weight (g/mol) 246.072
PubChem CID 5084102
Formula Weight 246.07
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.