Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%

Catalog No. p-3737308
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, C5H12N2S2, CAS Number-5108-96-3 | CAS: 5108-96-3 | C5H12N2S2 | 164.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5108-96-3
Molecular Formula C5H12N2S2
Molecular Weight (g/mol) 164.28
MDL Number MFCD00012720
InChI Key VSWDORGPIHIGNW-UHFFFAOYSA-M
Synonym ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium
PubChem CID 4311638
IUPAC Name pyrrolidine-1-carbodithioate
SMILES C1CCN(C1)C(=S)[S-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 150.0°C to 155.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol and chloroform
Formula Weight 164.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Pyrrolidinecarbodithioic acid, ammonium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 225-834-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.