Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-sec-Butyl-4-nitrobenzene 97.0+%, TCI America™
SDP

Catalog No. B242825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B242825G 25 g
B24285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B242825G Supplier TCI America Supplier No. B242825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4237-40-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00130025
InChI Key IQRMCUYGEZOTSV-UHFFFAOYNA-N
PubChem CID 107249
IUPAC Name 1-(butan-2-yl)-4-nitrobenzene
SMILES CCC(C)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Boiling Point 144°C
Quantity 25 g
Formula Weight 179.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-sec-Butyl-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.