Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butoxycarbonyl-3-cyanopiperidine 98.0+%, TCI America™
SDP

Catalog No. B45151G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 91419-53-3
Molecular Formula C11H18N2O2
Molecular Weight (g/mol) 210.277
MDL Number MFCD01861222
InChI Key UEFZTXGFHKPSFS-UHFFFAOYSA-N
Synonym 1-n-boc-3-cyanopiperidine, n-boc-3-cyanopiperidine, 1-boc-3-cyanopiperidine, +/--1-n-boc-3-cyano-piperidine, n-boc-3-cyano-piperidine, 1-n-boc-3-cyano-piperidine, 3-cyano-piperidine-1-carboxylic acid tert-butyl ester, 1-n-boc-piperidine-3-carbonitrile, 1-tert-butoxycarbonyl-3-cyanopiperidine, tert-butyl-3-cyanopiperidine-1-carboxylate
PubChem CID 2756785
IUPAC Name tert-butyl 3-cyanopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C#N

Specifications

Melting Point 67°C
Color White
Boiling Point 40°C
Quantity 1 g
UN Number 3439
Formula Weight 210.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butoxycarbonyl-3-cyanopiperidine
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.