Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(tert-Butoxycarbonyl)-3-pyrrolidinol 98.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 61°C |
Color | White-Yellow |
Quantity | 25 g |
Formula Weight | 187.24 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 1-(tert-Butoxycarbonyl)-3-pyrrolidinol |
Safety and Handling
TSCA | No |
Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction