Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butoxycarbonyl-4-(methylamino)piperidine 98.0+%, TCI America™
SDP

Catalog No. p-7123300
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 147539-41-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD02259411
InChI Key CZYUGTLMFHDODF-UHFFFAOYSA-N
Synonym 1-boc-4-methylaminopiperidine, tert-butyl 4-methylamino piperidine-1-carboxylate, 1-boc-4-methylamino piperidine, 1-boc-4-methylaminopieridine, n-boc-4-methylaminopieridine, n-boc-4-methylaminopiperidine, 4-methylamino-piperidine-1-carboxylic acid tert-butyl ester, 4-methylamino piperidine, n1-boc protected, n-boc-4-n-methylamino piperidine, 4-n-methylamino-1-boc-piperidine
PubChem CID 15380702
IUPAC Name tert-butyl 4-(methylamino)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)NC

Specifications

Color Colorless
Quantity 1 g
Formula Weight 214.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-tert-Butoxycarbonyl-4-(methylamino)piperidine
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.