Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-butyl) 2-methyl 1H-indole-1,2-dicarboxylate, 97%, Thermo Scientific™

Catalog No. CC27822DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC27822DA 1 g
CC27822CB 250 mg
CC27822DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC27822DA Supplier Thermo Scientific Chemicals Supplier No. CC27822DA
Add to Cart

Chemical Identifiers

CAS 163229-48-9
Molecular Formula C15H17NO4
Molecular Weight (g/mol) 275.304
InChI Key UJVYYFYNECMKRA-UHFFFAOYSA-N
Synonym 1-tert-butyl 2-methyl 1h-indole-1,2-dicarboxylate, 1-tert-butyl 2-methyl indole-1,2-dicarboxylate, 1-tert-butyl-2-methyl-1h-indole-1,2-dicarboxylate, 1h-indole-1,2-dicarboxylic acid 1-tert-butyl 2-methyl ester, 1h-indole-1,2-dicarboxylicacid, 1-1,1-dimethylethyl 2-methyl ester
PubChem CID 2795473
IUPAC Name 1-O-tert-butyl 2-O-methyl indole-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C2=CC=CC=C2C=C1C(=O)OC

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 275.3
Percent Purity 97%
Chemical Name or Material 1-(tert-Butyl) 2-methyl 1H-indole-1, 2-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.