Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trifluoromethanesulfonyl)imidazole 98.0+%, TCI America™
SDP

Catalog No. T31771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31771G 1 g
T31775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31771G Supplier TCI America Supplier No. T31771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29540-81-6
Molecular Formula C4H3F3N2O2S
Molecular Weight (g/mol) 200.14
MDL Number MFCD00037891
InChI Key YGABUCCNCBMODG-UHFFFAOYSA-N
PubChem CID 97300
IUPAC Name 1-trifluoromethanesulfonyl-1H-imidazole
SMILES FC(F)(F)S(=O)(=O)N1C=CN=C1

Specifications

Melting Point 19°C
Color Yellow
Boiling Point 40°C
Quantity 1 g
Formula Weight 200.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Trifluoromethanesulfonyl)imidazole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.