Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trifluoromethyl)vinyl acetate, 97%

Catalog No. p-7026233
Click to view available options
Quantity:
50 g
250 g

CAS: 2247-91-8 | C5H5F3O2 | 154.088 g/mol

1-(Trifluoromethyl)vinyl acetate is used as a chemical, organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trifluoromethyl)vinyl acetate is used as a chemical, organic Intermediates.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2247-91-8
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.088
MDL Number MFCD00040844
InChI Key VOKGSDIHTCTXDS-UHFFFAOYSA-N
Synonym 1-trifluoromethyl vinyl acetate, 1-trifluoromethyl ethenyl acetate, 1-propen-2-ol, 3,3,3-trifluoro-, acetate, 1-propen-2-ol, 3,3,3-trifluoro-, 2-acetate, trifluoroacetone enol acetate, trifluoroacetone enolacetate, alpha-trifluoromethylvinyl acetate, acmc-1cjli, 3,3,3-trifluoropropen-2-yl acetate, alpa-trifluoromethyl vinyl acetate
PubChem CID 75254
IUPAC Name 3,3,3-trifluoroprop-1-en-2-yl acetate
SMILES CC(=O)OC(=C)C(F)(F)F

Specifications

Density 1.2
Boiling Point 86°C to 88°C
Flash Point 2°C (35°F)
Odor Pungent
Refractive Index 1.342
Quantity 50 g
UN Number UN3272
Beilstein 2078331
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 154.09
Percent Purity 97%
Chemical Name or Material 1-(Trifluoromethyl)vinyl acetate
Show More Show Less
Hazard Category H225-H302+H312+H332-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 218-838-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.