Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Triisopropylsilyl)pyrrole 96.0+%, TCI America™
SDP

Catalog No. T167725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T167725ML 25 mL
T16775ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T167725ML Supplier TCI America Supplier No. T167725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87630-35-1
Molecular Formula C13H25NSi
Molecular Weight (g/mol) 223.44
MDL Number MFCD00054932
InChI Key FBQURXLBJJNDBX-UHFFFAOYSA-N
Synonym 1-triisopropylsilyl pyrrole, 1-triisopropylsilyl-1h-pyrrole, n-triisopropylsilylpyrrole, 1h-pyrrole, 1-tris 1-methylethyl silyl, tri propan-2-yl-pyrrol-1-ylsilane, 1-tris propan-2-yl silyl-1h-pyrrole, pubchem9194, 1-triisopropylsilylpyrrole, 1-triisoproylsilyl-pyrrole, acmc-1bkd7
PubChem CID 145136
IUPAC Name 1-[tris(propan-2-yl)silyl]-1H-pyrrole
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC=C1

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 223.44
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Triisopropylsilyl)pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.