Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Undecanol, 98%

Catalog No. p-210512
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle

CAS: 112-42-5 | C11H24O | 172.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-42-5
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00004751
InChI Key KJIOQYGWTQBHNH-UHFFFAOYSA-N
Synonym 1-undecanol, undecyl alcohol, undecanol, n-undecanol, n-undecyl alcohol, hendecanoic alcohol, hendecyl alcohol, 1-hendecanol, n-hendecylenic alcohol, tip-nip
PubChem CID 8184
ChEBI CHEBI:87499
IUPAC Name undecan-1-ol
SMILES CCCCCCCCCCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 11.0°C
Color Colorless to Yellow
Density 0.8310g/mL
Boiling Point 146.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4370 to 1.4420
Linear Formula CH3(CH2)10OH
Quantity 100 mL
Beilstein 01, 427
Specific Gravity 0.831
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in 60% alcohol
Formula Weight 172.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Undecanol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 203-970-5
RTECSNumber YQ3155000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.