Learn More
10,11-Dihydrocarbamazepine, 99%
Description
10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.
Solubility
Soluble in dichloromethane. Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 204°C to 206°C |
| Quantity | 250 mg |
| Beilstein | 196969 |
| Solubility Information | Soluble in dichloromethane. Insoluble in water. |
| Formula Weight | 238.29 |
| Percent Purity | 99% |
| Chemical Name or Material | 10,11-Dihydrocarbamazepine |
Safety and Handling
| EINECSNumber | 222-649-0 |
| RTECSNumber | HN8300000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.