Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 10-DEBC hydrochloride
GREENER_CHOICE

Catalog No. 255810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-581-0 10 mg
25-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-581-0 Supplier Tocris Bioscience™ Supplier No. 2558/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Akt/PKB inhibitor

Selective inhibitor of Akt/PKB. Inhibits IGF-1-stimulated phosphorylation and activation of Akt (complete inhibition at 2.5 μM), suppressing downstream activation of mTOR, p70 S6 kinase and S6 ribosomal protein. Shows no activity at PDK1, SGK1 or PI 3-kinase. Inhibits cell growth (IC50 ∽ 2-6 μM) and induces apoptosis in rhabdomyosarcoma cells.

Chemical Identifiers

CAS 925681-41-0
Molecular Formula C20H26Cl2N2O

Specifications

Quantity 10 mg
Formula Weight 381.34
Purity >99%
Molecular Weight (g/mol) 381.341
Chemical Name or Material 10-DEBC hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.