Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
10% Formalin in 5.2 mL Vial, Cenmed™

Click to view available options
Quantity:
500 x 5.2 mL
Description
- 10% concentration of formalin ensures effective fixation, maintaining tissue integrity for accurate histopathological analysis
- Packaged in a convenient 5.2 mL vial, this solution offers precise measurements, facilitating ease of use and enhancing accuracy in your experiments
- Trust in the 10% Formalin for dependable tissue fixation, setting a new standard for excellence in histopathology research
Specifications
Specifications
Physical Form | Liquid |
CAS | 50-00-0 |
Color | Clear |
Quantity | 500 x 5.2 mL |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction