Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-Methylphenothiazine 98.0+%, TCI America™
SDP

Catalog No. M251725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M251725G 25 g
M25175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M251725G Supplier TCI America Supplier No. M251725G

Chemical Identifiers

CAS 1207-72-3
Molecular Formula C13H11NS
Molecular Weight (g/mol) 213.30
MDL Number MFCD00041836
InChI Key QXBUYALKJGBACG-UHFFFAOYSA-N
Synonym 10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
PubChem CID 71015
IUPAC Name 10-methyl-10H-phenothiazine
SMILES CN1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 25 g
Formula Weight 213.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 10-Methylphenothiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.