Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-Phenylanthracene-9-boronic acid, 98%

Catalog No. AAH6491103
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6491103 1 g
AAH6491106 5 g
AAH64911MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6491103 Supplier Thermo Scientific Chemicals Supplier No. H6491103
Only null left
Add to Cart

CAS: 334658-75-2 | C20H15BO2 | 298.15 g/mol

It is used as pharmaceutical intermediate and as OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as OLED materials.

Solubility
Soluble in methanol.

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334658-75-2
Molecular Formula C20H15BO2
Molecular Weight (g/mol) 298.15
MDL Number MFCD11111989
InChI Key RVPCPPWNSMAZKR-UHFFFAOYSA-N
Synonym 10-phenylanthracen-9-yl boronic acid, 10-phenyl-9-anthracene boronic acid, 10-phenyl-9-anthraceneboronic acid, 10-phenylanthracene-9-boronic acid, 10-phenylantrhacen-9-yl boronic acid, boronic acid, 10-phenyl-9-anthracenyl, pubchem19639, 9-borono-10-phenylanthracene, 10-phenylanthracene-9-ylboronic acid, 10-phenyl-9-anthryl boronic acid
PubChem CID 22247164
IUPAC Name (10-phenylanthracen-9-yl)boronic acid
SMILES OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 11434760
Sensitivity Light sensitive
Solubility Information Soluble in methanol.
Formula Weight 298.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 10-Phenylanthracene-9-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.