Learn More
Thermo Scientific™ 100% Graphite Ferrules
Description
- Easy-to-use
- Form a soft grip with the column and provide a stable seal
Specifications
Specifications
| For Use With (Equipment) | For Thermo M4 Nut |
| Description | 0.53mm I.D. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.