Learn More
Invitrogen™ RSK2 Recombinant Rabbit Monoclonal Antibody (K01_2P36)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA564198
Description
RSK2 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
RPS6KA3 (RSK2) is a serine/threonine kinase that modulates many cellular processes including transcription, adhesion, growth, survival, and proliferation. RPS6KA3 contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in the gene have been associated with Coffin-Lowry syndrome (CLS).
Specifications
| RSK2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 | |
| P51812 | |
| RPS6KA3 | |
| A synthetic peptide of human Rsk 2/MAPKAP Kinase 1b. | |
| 100 μL | |
| Primary | |
| Human, Hamster | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| K01_2P36 | |
| Unconjugated | |
| RPS6KA3 | |
| 90 kDa ribosomal protein S6 kinase 3; C130006E23; CLS; HU-3; Insulin-stimulated protein kinase 1; ISPK1; ISPK-1; MAP kinase-activated protein kinase 1b; MAPK-activated protein kinase 1b; MAPKAP kinase 1b; MAPKAPK1B; MAPKAPK-1b; MPK-9; MRX19; p90-RSK 3; p90-RSK2; p90RSK3; pp90RSK2; RGD1563860; ribosomal protein S6 kinase 2; ribosomal protein S6 kinase A3; ribosomal protein S6 kinase alpha-3; ribosomal protein S6 kinase polypeptide 3; ribosomal protein S6 kinase, 90kDa, polypeptide 3; ribosomal S6 kinase 2; Rps6ka3; Rps6ka-rs1; RSK; Rsk2; RSK-2; S6K-alpha3; S6K-alpha-3 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 100750954, 6197 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.