Learn More
Invitrogen™ Zika Virus NS1 (strain H/PF/2013) Recombinant Rabbit Monoclonal Antibody (HL2567)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA556930

Description
Zika Virus NS1 (strain H/PF/2013) Recombinant Monoclonal Antibody for Western Blot
As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. & Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
Specifications
Zika Virus NS1 (strain H/PF/2013) | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Western Blot | |
HL2567 | |
Unconjugated | |
Zika NS1; ZIKV NS1; ZIKV-NS1 | |
Synthetic peptide of Zika virus NS1 protein. (strain: H/PF/2013 ). | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.