Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ mCherry Polyclonal Antibody

Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA5143576
Description
mCherry Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
Specifications
mCherry | |
Polyclonal | |
Unconjugated | |
mCherry | |
Full length recombinant protein expressed in and purified from E. coli. | |
RUO | |
Chemical | |
Antibody | |
IgY |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Conc. Not Determined | |
PBS with 0.02% sodium azide | |
Chicken | |
100 μL | |
Primary | |
4°C | |
Liquid | |
Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction