Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences 10B Chemically Competent Cells
SCI_ED

Catalog No. S000763
Change view
Click to view available options
Quantity:
12 x 50 μL
6 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000763 12 x 50 μL
S000764 6 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000763 Supplier G-Biosciences Supplier No. 7862027
Add to Cart
Add to Cart

G-Biosciences 10B Chemically Competent E. coli is a derivative of the popular DH10B.

It is T1 phage resistant and endA deficient for high-quality plasmid preparations. Chemically competent 10B E. coli cells suitable for high efficiency transformation in a wide variety of applications.

Specifications

Product Type Competent Cells
Sterility Sterile
Species E. coli
Format Vial
For Use With (Application) DNA Transformation
Quantity 12 x 50 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.