Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 10nm Streptavidin Gold Conjugate (10OD)
SDP

Catalog No. 2521000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2521000 1 mL
2520200 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2521000 Supplier Novus Biologicals™ Supplier No. 2521000

Streptavidin gold nanoparticle conjugates can be used in conjunction with biotinylated reagents such as biotinylated antibodies or proteins, in applications such as lateral flow immunoassays or electron microscopy.

InnovaCoat GOLD nanoparticles have a proprietary surface coating which covalently binds antibodies, proteins, peptides or any other biomolecule with free amine groups to form highly stable conjugates. The surface coating can withstand the most extreme conditions (e.g. 2.5M NaOH at 70°C for 2 hours), making InnovaCoat™ GOLD an ideal reagent for a wide range of applications.

Specifications

Quantity 1 mL
Content And Storage 4°C Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.