Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ 10X MOPS Concentrated Solution, OmniPur™, Calbiochem™,

Catalog No. 6350500ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-505-00ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 63-505-00ML Supplier MilliporeSigma™ Supplier No. 6350500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for RNA electrophoresis in agarose. Buffering range 6.5 to 7.9. Each liter of concentrate makes 10L of 1X MOPS. Prepared with MOPS and DEPC for RNA applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2 , 7732-18-5 , 6131-90-4 , 64-19-7 , 6381-92-6
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-n-morpholino propanesulfonic acid, 4-morpholinepropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-morpholinopropane-1-sulfonic acid, 3-n-morpholinopropanesulfonic acid, morpholinopropane sulfonic acid, 3-morpholinopropanesulfonic acid, unii-273bp63nv3, morpholinopropanesulfonic acid, 3-[N-Morpholino]propane sulfonic Acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Concentration 10X
For Use With (Application) Agarose Gel Electrophoresis, Polyacrylamide Gel Electrophoresis
Physical Form Solution
Quantity 500 mL
Solution Type Buffer
DNase None Detected
Protease None Detected
Solubility Information Soluble in water
Grade Molecular Biology
Chemical Name or Material 10X MOPS Concentrated Solution
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.