Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free

Catalog No. am9624
Encompass_Preferred
Click to view available options
Quantity:
500 mL
1 L

PBS (phosphate buffered saline) is a pH-adjusted blend of ultrapure-grade phosphate buffers and saline solutions.

PBS (phosphate buffered saline) is a pH-adjusted blend of ultrapure-grade phosphate buffers and saline solutions which, when diluted to a 1X working concentration, contains 137 mM NaCl, 2.7 mM KCl, 8 mM Na2HPO4, and 2 mM KH2PO4. Each 10X PBS solution is ready to use upon dilution to the desired concentration. This molecular biology-grade PBS is certified RNase-free; it is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
pH 7.4
Physical Form Liquid
Product Line Ambion™
Quantity 500 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.