Description
- 1 M TACS Safe-TdT™ Buffer
- 0.5mg/mL BSA (RIA Grade)
- 0.6mM 2-mercaptoethanesulfonic acid (MESNA)
Specifications
Specifications
| Quantity | 100 mL |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |