Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11,12-Dihydro-11-phenylindolo[2,3-a]carbazole 97.0+%, TCI America™
SDP

Catalog No. D48601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48601G 1 g
D4860200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48601G Supplier TCI America Supplier No. D48601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1024598-06-8
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.41
MDL Number MFCD22581305
InChI Key IENOQUQIVSMWGP-UHFFFAOYSA-N
Synonym 11-Phenylindolo[2,3-a]carbazole
PubChem CID 59365871
IUPAC Name 11-phenyl-11H,12H-indolo[2,3-a]carbazole
SMILES N1C2=CC=CC=C2C2=C1C1=C(C=C2)C2=C(C=CC=C2)N1C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 332.41
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 11,12-Dihydro-11-phenylindolo[2,3-a]carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.