Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11-Bromoundecanoic Acid 97.0+%, TCI America™
SDP

Catalog No. B0389100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0389100G 100 g
B038925G 25 g
B0389500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0389100G Supplier TCI America Supplier No. B0389100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2834-05-1
Molecular Formula C11H21BrO2
Molecular Weight (g/mol) 265.19
MDL Number MFCD00002732
InChI Key IUDGNRWYNOEIKF-UHFFFAOYSA-N
Synonym undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid
PubChem CID 17812
IUPAC Name 11-bromoundecanoic acid
SMILES C(CCCCCBr)CCCCC(=O)O

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 190°C
Quantity 100 g
Formula Weight 265.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 11-Bromoundecanoic Acid
EINECSNumber (2)-2652
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.